missing translation for 'onlineSavingsMsg'
Learn More

ARD1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 14803-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16813634

  • 319.00€ / 150 microlitros

Envío estimado: 21-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

N-alpha-acetylation is one of the most common protein modifications that occurs during protein synthesis and involves the transfer of an acetyl group from acetyl-coenzyme A to the protein alpha-amino group. ARD1A, together with NATH.
Especificaciones

Especificaciones

ARD1A
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P41227, Q9QY36
ARD1, ARD1A, DXS707, NAA10, TE2
Rabbit
IgG
150 μL
-20°C
Primary
363518, 56292, 8260
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
NAA10
Liquid
Naa10
ARD1A Fusion Protein Ag6519
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados