missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ AQP1 (Human) Recombinant Protein
Human AQP1 full-length ORF ( AAH22486, 1 a.a. - 269 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000358-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MASEFKKKLFWRAVVAEFLATTLFVFISIGSALGFKYPVGNNQTAVQDNVKVSLAFGLSIATLAQSVGHISGAHLNPAVTLGLLLSCQISIFRALMYIIAQCVGAIVATAILSGITSSLTGNSLGRNDLADGVNSGQGLGIEIIGTLQLVLCVLATTDRRRRDLGGSAPLAIGLSVALGHLLAIDYTGCGINPARSFGSAVITHNFSNHWIFWVGPFIGGALAVLIYDFILAPRSSDLTDRVKVWTSGQVEEYDLDADDINSRVEMKPK
Especificaciones
AAH22486 | |
aquaporin 1 (Colton blood group) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AQP1 | |
GST |
358 | |
Wheat germ expression system | |
25 μg | |
AQP-CHIP, CHIP28, CO, MGC26324 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |