missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Path-ID™ Multiplex One-Step Kit

The Path-ID™ Multiplex One-Step RT-PCR Kit is designed for multiplex, quantitative, reverse transcription PCR (qRT-PCR).

Marca:  Applied Biosystems™ 4428207

Detalles adicionales : Peso : 0.00450kg

 Ver más versiones de este producto

Código de producto. 15880383

  • 1210.00€ / 500 reacciones


En stock 9
Añadir a la cesta

Descripción

Descripción

Order Info

Built-In Hot Start. Guaranteed minimum shelf life is 60 days.

Especificación

Especificación

Primer-Probe Compatability: TaqMan™
Passive Reference Dye: ROX (Pre-mixed)
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
7mL of 2X Multiplex RT-PCR Buffer; 1.4mL of 10X Multiplex Enzyme Mix including AmpliTaq Gold and ArrayScript; 25mL of Nuclease-free water
Total RNA, Viral RNA, mRNA
Store the kit at -20°C; the Nuclease-free water may be stored at -20°C, 4°C, or room temperature.
500 reactions
Primer-Probe Detection
Real time PCR (qPCR)
Liquid
AmpliTaq Gold™ DNA Polymerase
Standard
500 Reactions
Videos

For Research Use Only. Not for use in diagnostic procedures.