missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apelin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA551279
Detalles adicionales : Peso : 0.01000kg
Descripción
The antibody detects endogenous levels of total Apelin protein.
Apelin Polyclonal antibody specifically detects Apelin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).Especificaciones
Apelin | |
Polyclonal | |
Unconjugated | |
Apln | |
6030430G11Rik; AGTRL1 ligand; APEL; Apelin; apelin, AGTRL1 ligand; Apelin-13; Apelin-28; Apelin-31; Apelin-36; APJ endogenous ligand; APLN; preproapelin; XNPEP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
30878, 58812, 8862 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9R0R3, Q9R0R4, Q9ULZ1 | |
Apln | |
Synthetic peptide of human APLN | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |