missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ AP3S1 (Human) Recombinant Protein
Human AP3S1 full-length ORF ( AAH00804.1, 1 a.a. - 193 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001176-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MIKAILIFNNHGKPRLSKFYQPYSEDTQQQIIRETFHLVSKRDENVCNFLEGGLLIGGSDNKLIYRHYATLYFVFCVDSSESELGILDLIQVFVETLDKCFENVCELDLIFHVDKVHNILAEMVMGGMVLETNMNEIVTQIDAQNKLEKSEAGLAGAPARAVSAVKNMNLPEIPRNINIGDISIKVPNLPSFK
Especificaciones
AAH00804.1 | |
adaptor-related protein complex 3, sigma 1 subunit | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AP3S1 | |
GST |
1176 | |
Wheat germ expression system | |
10 μg | |
CLAPS3, Sigma3A | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |