missing translation for 'onlineSavingsMsg'
Learn More

AP2 alpha Antibody (A6/2/2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NB600-202F

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216105

  • 424.40€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

AP2 alpha Monoclonal specifically detects AP2 alpha in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

AP2 alpha
Monoclonal
FITC
PBS with 0.05% Sodium Azide
TFAP2A
Peptide to C-Terminal of AP2 alpha coupled to KLH. PNSHTDNNAKSSDKEEKHRK
0.1 mL
Cancer, Transcription Factors and Regulators
7020
Human
Purified
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
A6/2/2
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha
Mouse
Protein A purified
RUO
Primary
This antibody detects both the alpha and beta forms of AP2.
Store at 4C in the dark.
IgG1
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales