missing translation for 'onlineSavingsMsg'
Learn More

AP1M2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10618-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817113

  • 109.00€ / 20 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a subunit of the heterotetrameric adaptor-related protein comlex 1 (AP-1), which belongs to the adaptor complexes medium subunits family. This protein is capable of interacting with tyrosine-based sorting signals. Alternative splicing results in multiple transcript variants.
Especificaciones

Especificaciones

AP1M2
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9WVP1, Q9Y6Q5
AP 1 complex subunit mu 2, AP mu chain family member mu1B, AP1 mu2, AP1M2, HSMU1B, MU 1B, Mu adaptin 2, MU1B, Mu1B adaptin, mu2
Rabbit
IgG
20 μL
-20°C
Primary
10053, 11768, 367038
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
AP1M2
Liquid
Ap1m2
AP1M2 Fusion Protein Ag0961
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados