missing translation for 'onlineSavingsMsg'
Learn More

Carga de antimonio, 1-3 mm, Puratronic™, 99,9999 % (base metálica), Thermo Scientific Chemicals

Catalog Number p-7048633
Click to view available options
Cantidad:
10 g
50 g
250 g
Este artículo no se puede devolver. Vea la política de devoluciones
7440-36-0
Sb
121.76
MFCD00011211 MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
5354495
CHEBI:30304
antimony
[Sb]
Este artículo no se puede devolver. Vea la política de devoluciones
7440-36-0
Sb
121.76
MFCD00011211 MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
5354495
CHEBI:30304
antimony
[Sb]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Antimony shot
Nombre de nota 1 to 3mm (0.04 to 0.1 in.)
Notas del porcentaje del ensayo (base metálica)
Cantidad 10 g
Porcentaje de pureza 99.9999%
Grado Puratronic™
Olor Odorless
Formulario Granalla

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'