missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XDH Polyclonal Antibody, HRP, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA128943
Detalles adicionales : Peso : 0.01000kg
Descripción
PA1-28943 detects Xanthine Oxidase from bovine samples.
XDH Polyclonal antibody specifically detects XDH in Bovine samples. It is validated for Immunomicroscopy, Western Blot.Especificaciones
XDH | |
Polyclonal | |
HRP | |
XDH | |
Unknown (protein for MGC:127098); xanthine dehydrogenase; Xanthine dehydrogenase/oxidase; xanthine oxidase; Xanthine oxidoreductase; XD; Xdh; xdh antibody; XDHA; xdha antibody; XO; XOR; xor antibody; Xox1; Xox-1 | |
Rabbit | |
IgG fraction | |
RUO | |
280960 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunomicroscopy, Western Blot | |
10 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2 | |
P80457 | |
XDH | |
Full length protein; Xanthine Oxidase from bovine buttermilk | |
1 mg | |
Primary | |
Bovine | |
Antibody | |
IgG |