Learn More
TTLL7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA531807
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: mouse brain. Predicted reactivity: Human (99%), Mouse (94%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TTLL7 Polyclonal antibody specifically detects TTLL7 in Human, Mouse samples. It is validated for Western Blot.Especificaciones
TTLL7 | |
Polyclonal | |
Unconjugated | |
TTLL7 | |
1110049N09Rik; 4921517B04Rik; C630030B20; mTTLL7; RP5-836J3.2; Testis development protein NYD-SP30; TTLL7; Tubulin polyglutamylase TTLL7; tubulin tyrosine ligase like 7; tubulin tyrosine ligase-like family member 7; tubulin tyrosine ligase-like family, member 7; tubulin--tyrosine ligase-like protein 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
70892, 79739 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.81 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
A4Q9F0, Q6ZT98 | |
TTLL7 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 204 of Human TTLL7 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |