Learn More
TNNT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA542109
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: WIHQLESEKF DLMAKLKQQK YEINVLYNRI SHAQKFRKGA GKGRVGGRWK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 85%
TNNT1 Polyclonal antibody specifically detects TNNT1 in Human, Mouse samples. It is validated for Western Blot.Especificaciones
TNNT1 | |
Polyclonal | |
Unconjugated | |
TNNT1 | |
ANM; AW146156; Fang2; MGC104241; NEM5; skeletal muscle slow-twitch TnT; slow skeletal muscle troponin T; ssTnT; sTnT; Tnnt1; TNT; TNTS; troponin T slow skeletal muscle type; troponin T slow type isoform sTnT1; troponin T slow type isoform sTnT2; troponin T type 1 (skeletal, slow); troponin T, slow skeletal muscle; troponin T1, skeletal, slow; troponin T1, slow skeletal type; troponin-T1, skeletal, slow; zgc:193831; zgc:193865 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
21955, 7138 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O88346, P13805 | |
TNNT1 | |
Synthetic peptide directed towards the middle region of human TNNT1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |