Learn More
TPM4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA530194
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: NT2D1, HeLa, mouse brain. Predicted reactivity: Mouse (96%), Rat (97%), Xenopus laevis (92%), Pig (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TPM4 Polyclonal antibody specifically detects TPM4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
TPM4 | |
Polyclonal | |
Unconjugated | |
TPM4 | |
2610528G24Rik; epididymis secretory protein Li 108; HEL-S-108; TM30p1; TM-4; TPM4; Tpm4.2; Tpm4.2cy; tropomycin 4; tropomyosin; tropomyosin 4; tropomyosin alpha-4 chain; Tropomyosin-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
24852, 326618, 7171 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P09495, P67936, Q6IRU2 | |
TPM4 | |
Recombinant protein fragment corresponding to a region within amino acids 54 and 226 of Human Tropomyosin 4 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |