missing translation for 'onlineSavingsMsg'
Learn More

TNF Rat anti-Mouse, PE-Cy7, Clone: MP6-XT22, BD

Rat Monoclonal Antibody

Marca:  BD Biosciences 557644

Detalles adicionales : Peso : 0.80000kg

 Ver más versiones de este producto

Código de producto. 12683356

  • 510.00€ / cada uno
Fecha estimada de envío: 22-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The MP6-XT22 antibody reacts with mouse tumor necrosis factor (TNF, also known as TNF-α). The immunogen used to generate this hybridoma was recombinant mouse TNF. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: MP6-XT22
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Recombinant mouse TNF

Intracellular Staining

Especificaciones

Especificaciones

TNF
Monoclonal
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Recombinant Mouse TNF
0.1 mg
Immunology
Mouse
IgG1
Flow Cytometry
MP6-XT22
PE-Cyanine7
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.