missing translation for 'onlineSavingsMsg'
Learn More

Stat4 (pY693) Mouse, Unlabeled, Clone: 38/P-STAT4, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 612738

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15826279

  • 470.00€ / cada uno
Fecha estimada de envío: 22-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The Stat proteins function both as cytoplasmic signal transducers and as activators of transcription. Seven mammalian Stat proteins have been identified: Stat1-4, Stat5a, 5b, and Stat6. Stat4 has been shown to play an important role in development of T helper cells, specifically the Th1 subset. Stat4 is activated by IL-12 and by type 1 interferons. Knockout mice supported the role that Stat4 plays in IL-12 signaling because lymphocytes from Stat 4-/- mice could not differentiate into Th1 cells or produce IFNγ in response to treatment with IL-12. IFNγ plays an important role in host defense. A key component in the activation of Stat4 is the phosphorylation on tyrosine and serine residues in response to IL-12 stimulation. IL-12 stimulation leads to the phosphorylation of Stat4 on tyrosine 693 and serine 721. Transcriptional activity of Stat4 has been shown to be significantly reduced when residues Y693 and S721 are mutated.

Western Blotting

Especificaciones

Especificaciones

Stat4 (pY693)
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Phosphorylated Human Stat4 (pY693)
50 μg
Cell Biology
Human, Mouse
IgG2b κ
Western Blot
38/P-STAT4
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.