Learn More
SPOP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA530546
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: A549, HCT116. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (95%), Xenopus laevis (97%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SPOP Polyclonal antibody specifically detects SPOP in Human samples. It is validated for Western Blot.Especificaciones
SPOP | |
Polyclonal | |
Unconjugated | |
SPOP | |
AI315626; BTBD32; HIB homolog 1; Pcif1; PDX-1 C-terminal-interacting factor 1; roadkill homolog 1; similar to speckle-type POZ protein; speckle type BTB/POZ protein; speckle type POZ protein; speckle-type POZ protein; Spop; SPOP1; TEF2; zgc:64140 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8405 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O43791 | |
SPOP | |
Recombinant fragment corresponding to a region within amino acids 139 and 354 of Human SPOP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |