Learn More
Brady™ Anti-skid Laminated Polyester Marking Tape
Product Code. 11761645 Shop All Brady Products
Change view
Click to view available options
Color:
Black and Yellow Striped
Yellow
Unit Size:
1 roll
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11761645 Black and Yellow Striped 1 roll
11315990 Yellow 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11761645 Supplier Brady™ Supplier No. 078147
  • ONLINE EXCLUSIVE PRICE
    103.00€
    99.69€ / 1 rollos
    Ahorro 3.31€
    3% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Anti-Skid (anti-slip) tapes help to prevent slipping and falling on wet or slippery floors. The rugged polyester tape is adhesive backed and coated with thousands of particles of durable aluminium oxide grit. Anti-Skid tape bonds permanently to clean, dry floors, withstands grease and oil and will work indoors or outdoors. Anti-Skid tape is available in a variety of colors and sizes to suit all of your floor marking needs
  • The rugged polyester tape is adhesive backed and coated with thousands of particles of durable aluminium oxide grit.
  • Anti-Skid tape bonds permanently to clean, dry floors, withstands grease and oil and will work indoors or outdoors.
  • Anti-Skid tape is available in a variety of colors and sizes to suit all of your floor marking needs.
Specifications
Alternate Part Number DP-STRIPED ANTI-SKID TAPE BLK/YEL
Certifications/Compliance ANSI A1264.2-2006, NSFI Certified for High Traction, OSHA 1910.144, OSHA 1910.22
Description Striped Anti-Skid Tape - Black / Yellow
Dimensions (L x W) 18.29 m x 50.80 mm
For Use With (Application) Traction on steps, slick floors, and egress pathway marking
Length (English) 60 ft.
Length (Metric) 18.29 m
Packaging Quantity 1 Roll
Temperature Range (English) -40°F to +158°F
Temperature Range (Metric) -40°C to +70°C
Thickness 0.162 mm
Width (English) 2 in.
Width (Metric) 50.8 mm
Adhesive Type Permanent Acrylic
Color Black and Yellow Striped
Material Polyester