missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STKLD1 Monoclonal Antibody (2B5), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA523149
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide Sequence: MLGPGSNRRR PTQGERGPGS PGEPMEKYQV LYQLNPGALG VNLVVEEMET KVKHVIKQVE CMDDHYASQA LEELMPLLKL RHAHISVYQE LFITWNGEI
STKLD1 Monoclonal antibody specifically detects STKLD1 in Human samples. It is validated for ELISA, Western Blot.Especificaciones
STKLD1 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q8NE28 | |
STKLD1 | |
STKL1 (NP_714921, 1 a.a. ∽ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
2B5 | |
Unconjugated | |
STKLD1 | |
C9orf96; Gm711; likely ortholog of H. sapiens chromosome 9 open reading frame 96 (C9orf96); LOC169436; probable inactive protein kinase-like protein SgK071; probable inactive protein kinase-like protein SgK071 homolog; probable protein kinase-like protein SgK071 homolog; RP11-244N20.8; serine/threonine kinase like domain containing 1; serine/threonine kinase-like domain containing 1; serine/threonine kinase-like domain-containing protein 1; Serine/threonine kinase-like domain-containing protein STKLD1; SGK071; Sk521; STKLD1; Sugen kinase 071 | |
Mouse | |
Affinity chromatography | |
RUO | |
169436 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |