missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF3B4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA518074
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology.
SF3B4 Polyclonal antibody specifically detects SF3B4 in Human samples. It is validated for Western Blot.Especificaciones
SF3B4 | |
Polyclonal | |
Unconjugated | |
Sf3b4 | |
49kD; AFD1; chunp6902; fb36a04; Hsh49; MGC10828; pre-mRNA-splicing factor SF3b 49 kDa subunit; SAP 49; Sap49; SAP49MGC10828; sf3b4; SF3b49; SF3b50; spliceosomal protein; spliceosome-associated protein (U2 snRNP); spliceosome-associated protein 49; Splicing factor 3B subunit 4; splicing factor 3b, subunit 4; splicing factor 3b, subunit 4, 49kD; splicing factor 3b, subunit 4, 49kDa; Unknown (protein for IMAGE:8120437); wu:fb36a04 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10262 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q15427 | |
Sf3b4 | |
Synthetic peptide sequence (AAGPISERNQDAT) corresponding to the N-terminus amino acids of SF3B4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |