missing translation for 'onlineSavingsMsg'
Learn More

SETMAR Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Marca:  Invitrogen PA518116

Detalles adicionales : Peso : 0.01000kg

Código de producto. 11386083

  • 473.00€ / 100 microgramos
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody is predicted to react with bovine based on sequence homology.

SETMAR Polyclonal antibody specifically detects SETMAR in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

SETMAR
Polyclonal
Unconjugated
SETMAR
5830404F24Rik; epidermal tumor expressed transcript 2; Etet2; Histone-lysine N-methyltransferase; Histone-lysine N-methyltransferase SETMAR; Mar1; METNASE; SET domain and mariner transposase fusion gene; SET domain and mariner transposase fusion gene-containing protein; SET domain and mariner transposase fusion gene-containing protein homolog; SET domain and mariner transposase fusion protein; SET domain and mariner transposase fusion protein homolog; SET domain without mariner transposase fusion; SET domain without mariner transposase fusion protein; Setmar; Transposon Hsmar1 transposase
Goat
Antigen affinity chromatography
RUO
6419
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q53H47
SETMAR
Synthetic peptide sequence (RWQKCVDCNGSYFD) corresponding to the C-terminus amino acids of SETMAR
100 μg
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales