missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Albumin Monoclonal Antibody (KT11), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA172509
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody can be used as a primary antibody for ELISA (1 μg/mL), or as a capture antibody in a sandwich ELISA (1-3 μg/mL).
Human Serum Albumin Monoclonal antibody specifically detects Human Serum Albumin in Human samples. It is validated for ELISA, Western Blot.Especificaciones
Human Serum Albumin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P02768 | |
ALB | |
Human albumin. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
KT11 | |
Unconjugated | |
ALB | |
Alb; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 | |
Mouse | |
Protein A | |
RUO | |
213 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |