missing translation for 'onlineSavingsMsg'
Learn More

SERPINC1 Polyclonal Antibody, Invitrogen™

Sheep Polyclonal Antibody

Marca:  Invitrogen PA143065

Detalles adicionales : Peso : 0.01000kg

Código de producto. 11508850

  • 273.00€ / 1 miligramo
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

PA1-43065 detects SERPINC1 in mouse samples. PA1-43065 has successfully been used in Western blot and ELISA applications. Predicted molecular weight of approximately 150 kDa. PA1-43065 was derived from mouse antithrombin.

SERPINC1 Polyclonal antibody specifically detects SERPINC1 in Mouse samples. It is validated for ELISA, Western Blot.
Especificaciones

Especificaciones

SERPINC1
Polyclonal
Unconjugated
Serpinc1
AI114908; antithrombin; Antithrombin 3; anti-thrombin 3; antithrombin III; antithrombin-III; AT3; At-3; AT3D; ATIII; hypothetical protein LOC540261; MGC22579; PRO0309; serine (or cysteine) peptidase inhibitor, clade C (antithrombin), member 1; serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1; serpin C1; serpin family C member 1; serpin peptidase inhibitor clade C member 1; serpin peptidase inhibitor, clade C (antithrombin), member 1; serpin peptidase inhibitor, clade C, member 1; Serpinc1; THPH7
Sheep
Ammonium sulfate precipitation, Ion-exchange chromatography
RUO
11905
-20° C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
9 mg/mL
50% water with 50% glycerol and no preservative
P32261
Serpinc1
Murine Antithrombin.
1 mg
Primary
Mouse
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales