missing translation for 'onlineSavingsMsg'
Learn More

SERPINA10 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA522100

Detalles adicionales : Peso : 0.01000kg

Código de producto. 13219538

  • 473.00€ / 100 microgramos
Fecha estimada de envío: 22-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Recommended positive controls: Molt-4. Predicted reactivity: Mouse (82%), Rat (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SERPINA10 Polyclonal antibody specifically detects SERPINA10 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

SERPINA10
Polyclonal
Unconjugated
SERPINA10
plasma protein associated with liver regeneration; protein Z-dependent protease inhibitor; PZ-dependent protease inhibitor; PZI; Rasp1; RASP-1; Regeneration-associated serpin 1; serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; serpin A10; serpin family A member 10; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10; serpin peptidase inhibitor, clade A, member 10; Serpina10; UNQ707/PRO1358; Zpi
Rabbit
Antigen affinity chromatography
RUO
51156
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9UK55
SERPINA10
Recombinant fragment corresponding to a region within amino acids 186 and 444 of SERPINA10 (Uniprot ID#Q9UK55)
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales