missing translation for 'onlineSavingsMsg'
Learn More

Angiotensinogen Polyclonal Antibody, Invitrogen™

Sheep Polyclonal Antibody

Marca:  Invitrogen PA547888

Detalles adicionales : Peso : 0.00010kg

Código de producto. 15528382

  • 422.00€ / 100 microgramos
Fecha estimada de envío: 08-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

In direct ELISAs, approximately 24% cross-reactivity with recombinant human Angiotensinogen is observed, and less than 1% cross-reactivity with recombinant mouse (rm) Serpin A1, rmSerpin C1, and rmSerpin F2 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

Angiotensinogen Polyclonal antibody specifically detects Angiotensinogen in Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Western Blot.
Especificaciones

Especificaciones

Angiotensinogen
Polyclonal
Unconjugated
AGT
AGT; AI265500; alpha-1 antiproteinase, antitrypsin; Ang; Ang I; Ang II; Ang III; Ang IV; AngI; AngII; Angiotensin 1-10; Angiotensin 1-4; Angiotensin 1-5; Angiotensin 1-7; Angiotensin 1-8; Angiotensin 1-9; Angiotensin 2-8; Angiotensin 3-8; Angiotensin I; angiotensin II; Angiotensin III; Angiotensin IV; angiotensin ll; Angiotensin-1; Angiotensin-2; Angiotensin-3; Angiotensin-4; Angiotensinogen; angiotensinogen (PAT); angiotensinogen (serpin peptidase inhibitor, clade A, member 8); ANHU; ANRT; Aogen; Des-Asp[1]-angiotensin II; PAT; pre-angiotensinogen; serine (or cysteine) proteinase inhibitor; serpin A8; serpin peptidase inhibitor, clade A, member 8; Serpina8
Sheep
Antigen affinity chromatography
RUO
11606, 24179
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Immunohistochemistry (Frozen), Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
P01015, P11859
AGT
CHO-derived recombinant mouse Serpin A8/Angiotensinogen Asp25-Val477
100 μg
Primary
Mouse, Rat
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales