missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARTS Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA519389
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody is predicted to react with bovine and canine based on sequence homology.
ARTS Polyclonal antibody specifically detects ARTS in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
ARTS | |
Polyclonal | |
Unconjugated | |
SEPTIN4 | |
4-Sep; apoptosis-related protein in the TGF-beta signaling pathway; ARTS; Bh5; BRADEION; Bradeion beta; brain protein H5; C17orf47; CE5B3; CE5B3 beta; cell division control-related protein 2; cell division control-related protein 2b; cerebral protein 7; EG3-1RVC; EG3RVC; expression gene 3 in rat visual cortex; expression gene 3-1 in rat visual cortex; H5; hCDCREL-2; hucep-7; MART; M-Septin; OTTMUSG00000001265; peanut-like 2; peanut-like 2 homolog; peanut-like protein 2; PNUTL2; SEP4; Sept4; septin 4; septin H5; Septin4; septin-4; septin-M; Uncharacterized protein C17orf47 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
5414 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O43236 | |
SEPTIN4 | |
Synthetic peptide sequence (RNKLTRESGTD) corresponding to the internal amino acids of 38231 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |