missing translation for 'onlineSavingsMsg'
Learn More

SEC22B Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen OSS00040W

Detalles adicionales : Peso : 0.01000kg

Código de producto. 11581292

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Glycerol (1:1) may be added for additional stability. Reconstitute in 100 μL of sterile water.

The protein encoded by this gene is a member of the SEC22 family of vesicle trafficking proteins. It seems to complex with SNARE and it is thought to play a role in the ER-Golgi protein trafficking. This protein has strong similarity to Mus musculus and Cricetulus griseus proteins.
Especificaciones

Especificaciones

SEC22B
Polyclonal
SEC22B
SEC22 vesicle-trafficking protein homolog B, SEC22 vesicle-trafficking protein-like 1, ERS24, ERS-24, Vesicle-trafficking protein SEC22b, SEC22L1, SEC
Rabbit
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
O75396, O08547, Q4KM74
SEC22B
A synthetic peptide from human Vesicle-trafficking protein SEC22b
RUO
20333, 310710, 9554
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales