Learn More
SARS Coronavirus Spike Protein Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA141513
Detalles adicionales : Peso : 0.01000kg
Descripción
The immunogen is 87% (13/15) homologous to SARS-COV-2 (COVID-19) Spike protein. The cross reactivity is not empirically tested. Suggested positive control: transfected mouse melanoma cell lysate.
SARS Coronavirus Spike Protein Polyclonal antibody specifically detects SARS Coronavirus Spike Protein in Virus samples. It is validated for Western Blot.Especificaciones
SARS Coronavirus Spike Protein | |
Polyclonal | |
Unconjugated | |
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
Synthetic peptide corresponding to residues 755-769 of the putative SARS Spike glycoprotein. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.2% gelatin and 0.05% sodium azide | |
Rabbit | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG |