missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgG2b (Heavy chain) Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marca: Invitrogen MA516819
Detalles adicionales : Peso : 0.01000kg
Descripción
Mouse anti Rat IgG2b Heavy Chain antibody, clone MARG2b-3 recognizes the gamma-2b heavy chain of rat immunoglobulin.
Rat IgG2b (Heavy chain) Monoclonal antibody specifically detects Rat IgG2b (Heavy chain) in Rat samples. It is validated for ELISA.Especificaciones
Rat IgG2b (Heavy chain) | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Rat IgG2b | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
ELISA | |
MARG2b-3 | |
Unconjugated | |
Mouse | |
Affinity chromatography | |
RUO | |
Rat | |
Liquid |