Learn More
Mouse anti-Rat IgG2a (Heavy chain) Secondary Antibody, HRP, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marca:  Invitrogen MA516818
734.70 EUR hasta el 2024-03-29
Use el código promocional "21615" para aplicar la oferta.
Detalles adicionales : Peso : 0.01000kg
Código de producto. 15286059
  • 1102.00€ / 500 microgramos
Fecha estimada de envío: 23-04-2024
para ver el stock
Descripción
Descripción
Mouse anti Rat IgG2a heavy chain antibody, clone MARG2a-1 recognizes the rat gamma 2a immunoglobulin heavy chain, and does not cross-react with other immunoglobulin classes or subclasses.
Rat IgG2a (Heavy chain) Monoclonal antibody specifically detects Rat IgG2a (Heavy chain) in Rat samples. It is validated for ELISA.
Especificaciones
Especificaciones
Rat IgG2a (Heavy chain)
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.01% thimerosal
Rat IgG2a
500 μg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG1
ELISA
MARG2a-1
HRP
Mouse
Affinity chromatography
RUO
Rat