Learn More
Mouse IgG1 Isotype Control (W3/25), Invitrogen™
Mouse Isotype Control
Marca: Invitrogen MA133300
Detalles adicionales : Peso : 0.01000kg
Descripción
Thermo Scientific Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
MA1-33300 detects IgG1 from rat samples.MA1-33300 has been successfully used in immunohistochemistry (frozen tissue) applications.Especificaciones
Mouse IgG1 | |
W3/25 | |
Unconjugated | |
Proprietary immunogen - Contact ABR for details. | |
100 Tests | |
Isotype Control (Primary) | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen) | |
0.1mg/mL | |
Mouse | |
Protein A | |
RUO | |
Rat | |
Liquid |