Learn More
Mouse anti-Rat IgE Secondary Antibody, FITC, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marca: Invitrogen MA516812
Detalles adicionales : Peso : 0.01000kg
Descripción
For FACS analysis, use 50 μL of the suggested working dilution to label 2x10^7 cells in 100 μL. Mouse anti Rat IgE antibody, clone MARE-1 recognizes rat epsilon heavy chain of immunoglobulin IgE and does not cross-react with other classes of rat immunoglobulin.
Rat IgE Monoclonal antibody specifically detects Rat IgE in Rat samples. It is validated for Flow Cytometry.Especificaciones
Rat IgE | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide | |
Rat IgE myeloma proteins (IR162, IR1016, IR2 and IR410) | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG1 |
Flow Cytometry | |
MARE-1 | |
FITC | |
Mouse | |
Affinity chromatography | |
RUO | |
Rat | |
Liquid |