Learn More
c-Raf Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA529333
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (96%), Rat (97%), Chicken (92%), Rhesus Monkey (100%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
c-Raf Polyclonal antibody specifically detects c-Raf in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
c-Raf | |
Polyclonal | |
Unconjugated | |
Raf1 | |
6430402F14Rik; AA990557; BB129353; CMD1NN; CRAF; C-RAF; C-Raf proto-oncogene, serine/threonine kinase; Craf1; Craf1 transforming; D830050J10Rik; EC 2.7.11.1; kinase Raf1; murine leukemia viral (v-raf-1) oncogene homolog 1 (3611-MSV); murine sarcoma 3611 oncogene 1; NS5; Oncogene RAF1; OTTHUMP00000207813; OTTHUMP00000209389; proto-oncogene c-RAF; RAF; raf proto-oncogene serine/threonine protein kinase; RAF proto-oncogene serine/threonine-protein kinase; RAF1; Raf-1; Raf-1 proto-oncogene, serine/threonine kinase; sarcoma 3611 oncogene; v-Raf; v-raf-1 leukemia viral oncogene 1; vraf1 murine le; v-raf-1 murine leukemia viral oncogene homolog 1; v-raf-1 murine leukemia viral oncogene-like protein 1; v-raf-leukemia viral oncogene 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
110157, 5894 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P04049, Q99N57 | |
Raf1 | |
Recombinant fragment corresponding to a region within amino acids 51 and 377 of Human RAF1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |