missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPY4R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA534106
Detalles adicionales : Peso : 0.01000kg
Descripción
Percent identity with other species by BLAST analysis: Human, Gibbon (100%) Gorilla, Panda (94%) Monkey, Bovine, Horse, Pig (88%) Bat, Rabbit (82%).
NPY4R Polyclonal antibody specifically detects NPY4R in Human samples. It is validated for Immunohistochemistry (Paraffin).Especificaciones
NPY4R | |
Polyclonal | |
Unconjugated | |
NPY4R | |
neuropeptide Y receptor type 4; neuropeptide Y receptor Y4; NPY4R; NPY4-R; NPYR-D; NYYR-D; Pancreatic polypeptide receptor 1; PP1; PPYR1; Y4; Y4 receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5540 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P50391 | |
NPY4R | |
Synthetic 17 amino acid peptide from C-terminus of human PPYR1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |