missing translation for 'onlineSavingsMsg'
Learn More

PPP4C Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA530128

Detalles adicionales : Peso : 0.01000kg

Código de producto. 13464667

  • 473.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Recommended positive controls: HepG2, NIH-3T3, JC, BCL-1. Predicted reactivity: Mouse (100%), Rat (99%), Xenopus laevis (99%), Rabbit (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PPP4C Polyclonal antibody specifically detects PPP4C in Human, Mouse samples. It is validated for ChIP Assay, Immunocytochemistry, Western Blot.
Especificaciones

Especificaciones

PPP4C
Polyclonal
Unconjugated
PPP4C
1110002D08Rik; AU016079; P PPX; pp4; PP4C; PPH3; Ppp4; PPP4C; PPX; PP-X; protein phosphatase 4 (formerly X), catalytic subunit; protein phosphatase 4 catalytic subunit; protein phosphatase 4, catalytic subunit; protein phosphatase X; protein phosphatase X, catalytic subunit; serine/threonine-protein phosphatase 4 catalytic subunit
Rabbit
Antigen affinity chromatography
RUO
5531, 56420
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ChIP Assay, Immunocytochemistry, Western Blot
0.29 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P60510, P97470
PPP4C
Recombinant fragment corresponding to a region within amino acids 1 and 275 of Human PPP4C
100 μL
Primary
Human, Mouse
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales