missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP2A alpha Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA515577
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody is predicted to react with bovine, porcine and rabbit based on sequence homology.
PP2A alpha Polyclonal antibody specifically detects PP2A alpha in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
PP2A alpha | |
Polyclonal | |
Unconjugated | |
PPP2CA | |
B55-GAMMA; hypothetical protein; IMYPNO; IMYPNO1; Methyl-PPP2CA; MGC33570; PP2A; PP2A alpha; PP2A C terminal; PP2A L9; PP2A Leucine 9; Pp2a1; PP2A-alpha; PP2Ac; PP2CA; PP2Calpha; Ppp2ca; PR52; PR55G; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2 catalytic subunit alpha; protein phosphatase 2, catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2,catalytic subunit alpha isozyme; protein phosphatase 2A alpha subunit; protein phosphatase 2A catalytic subunit alpha isoform; protein phosphatase 2a, catalytic subunit, alpha isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A; protein phosphatase-2A-alpha; R75353; replication protein C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; unnamed protein product | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
5515 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P67775 | |
PPP2CA | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PP2A alpha | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |