missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POTEF Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA546592
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: KVPRKDLIVM LRDTDVNKQD KQKRTALHLA SANGNSEVVK LLLDRRCQLN Sequence homology: Human: 100%
The function of this protein remains unknown.Especificaciones
POTEF | |
Polyclonal | |
Unconjugated | |
POTEF | |
POTEF | |
Synthetic peptide directed towards the middle region of human POTEF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A5A3E0 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
728378 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |