missing translation for 'onlineSavingsMsg'
Learn More

KV1.4 (KCNA4) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA112655

Detalles adicionales : Peso : 0.01000kg

Código de producto. 11566042

  • 473.00€ / 100 microgramos
Fecha estimada de envío: 08-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

KV1.4 (KCNA4) Polyclonal antibody specifically detects KV1.4 (KCNA4) in Human, Mouse, Rat samples. It is validated for Western Blot.
Especificaciones

Especificaciones

KV1.4 (KCNA4)
Polyclonal
PBS with 0.08% sodium azide
P15385, P22459, Q61423
KCNA4
Synthetic peptide, conjugated to KLH, corresponding to the N-terminus of the rat Kv14 potassium channel.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot
Unconjugated
KCNA4
cardiac potassium channel; fetal skeletal muscle potassium channel; HBK4; HK1; HPCN2; HUKII; KCHAN; Kcna4; KCNA4L; KCNA8; Kv1.4; Kv4; PCN2; potassium channel 2; potassium channel, voltage gated shaker related subfamily A, member 4; potassium voltage gated channel, shaker related subfamily, member 4; potassium voltage-gated channel subfamily A member 4; potassium voltage-gated channel, shaker-related subfamily, member 4; rapidly inactivating potassium channel; RCK4; RHK1; RK3; shaker-related potassium channel Kv1.4; type A potassium channel; voltage-gated K(+) channel HuKII; Voltage-gated potassium channel HBK4; voltage-gated potassium channel HK1; Voltage-gated potassium channel subunit Kv1.4
Rabbit
Ammonium sulfate precipitation
RUO
16492, 25469, 3739
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales