Learn More
POLR2I Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA541120
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: ACRNCDYQQE ADNSCIYVNK ITHEVDELTQ IIADVSQDPT LPRTEDHPCQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 83%; Zebrafish: 93%
POLR2I Polyclonal antibody specifically detects POLR2I in Human samples. It is validated for Western Blot.Especificaciones
POLR2I | |
Polyclonal | |
Unconjugated | |
POLR2I | |
2810002B19Rik; DNA-directed RNA polymerase II 14.5 kDa polypeptide; DNA-directed RNA polymerase II subunit I; DNA-directed RNA polymerase II subunit RPB9; hRPB14.5; Polr2i; polymerase (RNA) II (DNA directed) polypeptide I; polymerase (RNA) II (DNA directed) polypeptide I, 14.5kDa; polymerase (RNA) II subunit I; RNA polymerase II 14.5 kDa subunit; RNA polymerase II subunit B9; RNA polymerase II subunit I; RPB14.5; RPB9; Unknown (protein for MGC:133488); zgc:103515 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5438 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P36954 | |
POLR2I | |
Synthetic peptide directed towards the N-terminal of human POLR2I | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |