missing translation for 'onlineSavingsMsg'
Learn More

PM20D2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA535048

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15317464

  • 338.00€ / 400 microlitros


Fecha de envío estimada
del proveedor 10-06-2020
Añadir a la cesta

Descripción

Descripción

PM20D2 belongs to the peptidase M20A family. The function of PM20D2 remains unknown.
Especificaciones

Especificaciones

PM20D2
Polyclonal
PBS with 0.09% sodium azide
Q8IYS1
PM20D2; ACY1L2; Peptidase M20 domain-containing protein 2; Aminoacylase-1-like protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
135293
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PM20D2
Liquid
PM20D2
KLH conjugated synthetic peptide between 396-425 amino acids from the C-terminal region of human PM20D2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Videos
Documentos