missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin A2 Monoclonal Antibody (583603), Invitrogen™
Rat Monoclonal Antibody
Marca: Invitrogen MA524263
Detalles adicionales : Peso : 0.00010kg
Descripción
In direct ELISAs, no cross-reactivity with recombinant human Plexin A4, B1, B2, D1, recombinant mouse Plexin A1, A3, B3, or C1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Plexin A2 Monoclonal antibody specifically detects Plexin A2 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Especificaciones
Plexin A2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O75051, P70207 | |
PLXNA2 | |
CHO-derived recombinant mouse Plexin A2 Met35-Pro1237 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
Immunocytochemistry, Western Blot | |
583603 | |
Unconjugated | |
PLXNA2 | |
2810428A13Rik; AA589422; AW457381; FLJ11751; FLJ30634; KIAA0463; LOW QUALITY PROTEIN: plexin-A2; mKIAA0463; OCT; plex 2; PlexA2; plexin 2; plexin A2; plexin-2; plexin-A2; PLXN2; Plxna2; Semaphorin receptor OCT; si:dkey-164k14.1; transmembrane protein OCT; UNQ209/PRO235 | |
Rat | |
Protein A/G | |
RUO | |
18845, 5362 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |