Learn More
PLEKHH2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA543998
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: WQLLALCVGL FLPHHPFLWL LRLHLKRNAD SRTEFGKYAI YCQRCVERTQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%
PLEKHH2 Polyclonal antibody specifically detects PLEKHH2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Especificaciones
PLEKHH2 | |
Polyclonal | |
Unconjugated | |
PLEKHH2 | |
AI256725; E030001K05; KIAA2028; mKIAA2028; pleckstrin homology domain containing, family H (with MyTH4 domain) member 2; pleckstrin homology domain-containing family H member 2; pleckstrin homology, MyTH4 and FERM domain containing H2; PLEKHH1L; PLEKHH2; RGD1304935; si:ch73-105b23.3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
130271 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IVE3 | |
PLEKHH2 | |
Synthetic peptide directed towards the C-terminal of human PLEKHH2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |