missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC mu Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA130286
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: Mouse kidney lysate. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PKC mu Polyclonal antibody specifically detects PKC mu in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
PKC mu | |
Polyclonal | |
Unconjugated | |
Prkd2 | |
4930557O20Rik; 5730497N19Rik; AI325941; DKFZP586E0820; EPK2; HSPC187; nPKC-D1; nPKC-D2; nPKC-mu; nPKC-nu; PKC mu; PKC nu; PKC u; PKC v; Pkcm; PKCmu; PKC-MU; Pkcnu; PKC-NU; PKCu; PKC-u; PKCv; PKC-v; Pkd; PKD1; PKD2; PKD3; Prkcm; PRKCN; Prkd1; PRKD2; PRKD3; Protein kinase C mu type; protein kinase C nu type; protein kinase C, mu; protein kinase C, nu; protein kinase D; protein kinase D1; protein kinase D2; protein kinase D3; protein kinase EPK2; RGD1308054; Serine/threonine-protein kinase D1; serine/threonine-protein kinase D2; Serine/threonine-protein kinase D3 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
101540, 18760, 23683, 25865, 5587, 75292 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
O94806, Q15139, Q62101, Q8BZ03, Q8K1Y2, Q9BZL6 | |
PRKD1, Prkd2, Prkd3 | |
KLH conjugated synthetic peptide between 361-389 amino acids from the N-terminal region of human PKC mu | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |