missing translation for 'onlineSavingsMsg'
Learn More

PJCG2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA539862

Detalles adicionales : Peso : 0.00900kg

Código de producto. 15526731

  • 339.00€ / 100 microgramos


Fecha de envío estimada
del proveedor 26-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

PJCG2
Polyclonal
Unconjugated
OR7E13P
Olfactory-like receptor PJCG2
Rabbit
IgG
100 μg
-20°C
Primary
10820
Immunocytochemistry, Immunofluorescence
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Liquid
OR7E13P
A synthetic peptide derived from the internal region of human PJCG2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Videos
Documentos