missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP4K2C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA531550
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: U87-MG, MCF-7. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PIP4K2C Polyclonal antibody specifically detects PIP4K2C in Human samples. It is validated for Immunocytochemistry, Western Blot.Especificaciones
PIP4K2C | |
Polyclonal | |
Unconjugated | |
PIP4K2C | |
phosphatidylinositol 4-phosphate 5-kinase; phosphatidylinositol 5-phosphate 4-kinase type II gamma; Phosphatidylinositol 5-phosphate 4-kinase type-2 gamma; phosphatidylinositol-4-phosphate 5-kinase, type II, gamma; phosphatidylinositol-5-phosphate 4-kinase type 2 gamma; phosphatidylinositol-5-phosphate 4-kinase type-2 gamma; phosphatidylinositol-5-phosphate 4-kinase, type II, gamma; PI(5)P 4-kinase type II gamma; Pip4k2c; PIP4KII-gamma; PIP5K2C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79837 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8TBX8 | |
PIP4K2C | |
Synthetic peptide corresponding to a region within amino acids 78 and 170 of Human PIP5K2 gamma | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |