missing translation for 'onlineSavingsMsg'
Learn More

PIBF1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA530387

Detalles adicionales : Peso : 0.01000kg

Código de producto. 13494877

  • 473.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Recommended positive controls: HCT116. Predicted reactivity: Human (99%), Mouse (92%), Rat (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PIBF1 Polyclonal antibody specifically detects PIBF1 in Human samples. It is validated for Immunocytochemistry, Western Blot.
Especificaciones

Especificaciones

PIBF1
Polyclonal
Unconjugated
PIBF1
C13orf24; Centrosomal protein of 90 kDa; CEP90; PIBF; PIBF1; progesterone immunomodulatory binding factor 1; progesterone-induced blocking factor 1; progesterone-induced-blocking factor 1; RGD1305077; RP11-505F3.1
Rabbit
Antigen affinity chromatography
RUO
10464
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q8WXW3
PIBF1
Recombinant protein fragment corresponding to a region within amino acids 462 and 662 of Human PIBF1
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales