Learn More
PIK3CA Rabbit anti-Bovine, Human, Mouse, Rat, Clone: H.843.0, Invitrogen™
Rabbit Monoclonal Antibody
Marca:  Invitrogen MA514870
Detalles adicionales : Peso : 0.01000kg
Código de producto. 11524841
  • 363.00€ / 100 microlitros
Fecha de envío estimada
del proveedor 06-07-2020
Añadir a la cesta
Descripción
Descripción
Phosphatidylinositol 3-kinase is composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. The protein encoded by this gene represents the catalytic subunit, which uses ATP to phosphorylate PtdIns, PtdIns4P and PtdIns(4,5)P2. This gene has been found to be oncogenic and has been implicated in cervical cancers.
Especificaciones
Especificaciones
PIK3CA
Monoclonal
Unconjugated
PIK3CA
Liquid
PIK3CA
Synthetic peptide surrounding Asp520 of the sequence of human PI3K p110a
Affinity chromatography
RUO
Antibody
Monoclonal
Bovine, Human, Mouse, Rat
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
H.843.0
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
P32871, P42336, P42337, A0A0G2K344
PI3-kinase p110 subunit alpha; PI3-kinase subunit alpha; PI3K-alpha; PtdIns-3-kinase p110; phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha; phosphatidylinositol 3-kinase, catalytic, alpha polypeptide; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform; phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform; ptdIns-3-kinase subunit alpha; ptdIns-3-kinase sub
Rabbit
IgG
100 μL
-20°C
Primary
170911, 18706, 282306, 5290