missing translation for 'onlineSavingsMsg'
Learn More

STAT1 Recombinant Polyclonal Antibody (4HCLC), Invitrogen™

Rabbit Recombinant Polyclonal Antibody

Marca:  Invitrogen 710078

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15317174

  • 461.00€ / 100 microgramos
Fecha estimada de envío: 16-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.

STAT1 Recombinant Polyclonal antibody specifically detects STAT1 in Human samples. It is validated for ChIP Assay, ELISA, Immunocytochemistry, Western Blot.
Especificaciones

Especificaciones

STAT1
Recombinant Polyclonal
0.5 mg/mL
PBS with 0.09% sodium azide
P42224
STAT1
Recombinant protein corresponding to amino acids 134–218 of human STAT1
100 μg
Primary
Human
Antibody
IgG
ChIP Assay, ELISA, Immunocytochemistry, Western Blot
4HCLC
Unconjugated
STAT1
2010005J02Rik; AA408197; CANDF7; DD6G4-4; DKFZp686B04100; IMD31A; IMD31B; IMD31C; ISGF-3; LOW QUALITY PROTEIN: signal transducer and activator of transcription 1-alpha/beta; OTTHUMP00000165046; OTTHUMP00000205845; signal transducer and activator of transcription 1; signal transducer and activator of transcription 1, 91kD; signal transducer and activator of transcription 1, 91kDa; signal transducer and activator of transcription 1-alpha/beta; signal transducer and activator of transcription-1; STAT; STAT 1; STAT1; STAT-1; stat1 alpha; STAT91; transcription factor ISGF-3; transcription factor ISGF-3 components p91/p84
Rabbit
Protein A
RUO
6772
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales