missing translation for 'onlineSavingsMsg'
Learn More

Phospho-PP1 alpha (Thr320) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA517819

Detalles adicionales : Peso : 0.01000kg

Código de producto. 11597060

  • 467.00€ / 100 microlitros
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

It is not recommended to aliquot this antibody.

Phospho-PP1 alpha (Thr320) Polyclonal antibody specifically detects Phospho-PP1 alpha (Thr320) in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Especificaciones

Especificaciones

Phospho-PP1 alpha (Thr320)
Polyclonal
Unconjugated
PPP1CA
96A; CG6593; CG6593-PA; CG6593-PB; dism2; Dmel\CG6593; Dmel_CG6593; DmPp1-96A; GADD34; LD14639p; MGC15877; MGC1674; PP1; PP1 96A; PP1 alpha96a; Pp1-96A; PP1A; PP-1A; PP1A-96A; PP1alpha; PP1alpha[[1]]; PP1alpha96A; pp1alpha-96A; Pp1alpha-96A-PA; Pp1alpha-96A-PB; PP1c; PP1c 96A; PPP1A; Ppp1c; Ppp1ca; protein phosphatase 1; protein phosphatase 1 catalytic subunit alpha; protein phosphatase 1, catalytic subunit, alpha isoform; protein phosphatase 1, catalytic subunit, alpha isozyme; Protein phosphatase 1alpha at 96A; protein phosphatase type 1 alpha catalytic subunit; Protein phosphatase type 1 alpha, catalytic subunit; protein phosphatase-1d; serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit; Serine/threonine-protein phosphatase alpha-1 isoform; Serine/threonine-protein phosphatase PP1-alpha catalytic subunit; testicular tissue protein Li 155
Rabbit
Antigen affinity chromatography
RUO
19045, 24668, 5499
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
165 μg/mL
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
P62136, P62137, P62138
PPP1CA
Synthetic phosphopeptide corresponding to residues surrounding pThr320 of human PP1a
100 μL
Primary
Human, Mouse, Non-human Primate, Rat
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales