missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PP1 alpha (Thr320) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA538391
Detalles adicionales : Peso : 0.00900kg
Descripción
The protein encoded by this gene is one of the three catalytic subunits of protein phosphatase 1 (PP1). PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. Increased PP1 activity has been observed in the end stage of heart failure. Studies in both human and mice suggest that PP1 is an important regulator of cardiac function. Mouse studies also suggest that PP1 functions as a suppressor of learning and memory. Three alternatively spliced transcript variants encoding different isoforms have been found for this gene.Especificaciones
Phospho-PP1 alpha (Thr320) | |
Polyclonal | |
Unconjugated | |
PPP1CA | |
PP-1A, PP1a, MGC1674, MGC15877, PP1alpha, PPP1CA, Serine/threonine-protein phosphatase PP1-alpha catalytic subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19045, 24668, 5499 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P62136, P62137, P62138 | |
PPP1CA | |
A synthetic phosphopeptide derived from human PP1-alpha around the phosphorylation site of Thr320 (P-I-TP-P-P) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |