missing translation for 'onlineSavingsMsg'
Learn More

Phospho-PLCG2 (Tyr753) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA538244

Detalles adicionales : Peso : 0.00900kg

Código de producto. 15535171

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Phosphoinositide-specific phospholipase C (PLC) plays a crucial role in the initiation of receptor mediated signal transduction through the generation of the two second messengers, inositol 1,4,5-triphosphate and diacylglycerol from phosphatidylinositol 4,5-bisphosphate. There are many mammalian PLC isozymes, including PLC beta1, PLC beta2, PLC beta3, PLC beta4, PLC gamma1, PLC gamma2, PLC delta1, PLC delta2 and PLCe. PLC delta exists as four different isoforms. PLC delta1, a calcium signal amplifier, is activated by an atypical GTP-binding protein. In addition, PLC delta1 is an effector for GTP-binding protein transglutaminase II-mediated oxytocin receptor and alpha1B-adrenoreceptor signaling. Mouse PLC delta1 is highly expressed in brain, heart, lung and testis. PLC delta is abnormally accumulated in autopsied brains with Alzheimer′s disease (AD), suggesting that it may play a role in the pathology of AD. PLC delta2 is markedly expressed in type II intestinal metaplasia and in the adenocarcinoma. When PLC delta2 is expressed in type I intestinal metaplasia, the metaplasia is generally considered benigt, yet evolves toward neoplastic transformation. Thus, PLC delta2 expression may be a possible marker of gastric maligt transformation.
Especificaciones

Especificaciones

Phospho-PLCG2 (Tyr753)
Polyclonal
Unconjugated
PLCG2
1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma-2, Phosphoinositide phospholipase C-gamma-2, Phospholipase C-IV, PLC-IV, Phospholipase C-gamma-2, PLC-gamma-2, PLCG2, PLC gamma2, PLC- gamma2, PLC gamma-2
Rabbit
Antigen affinity chromatography
RUO
234779, 29337, 5336
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
P16885, Q8CIH5, P24135
PLCG2
A synthetic phosphopeptide derived from human PLCG2 around the phosphorylation site of Tyr753 (S-L-YP-D-V)
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales