missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-p47phox (Ser359) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA537807
Detalles adicionales : Peso : 0.00910kg
Descripción
A suggested positive control for Western blot is Hela cells.
Phospho-p47phox (Ser359) Polyclonal antibody specifically detects Phospho-p47phox (Ser359) in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
Phospho-p47phox (Ser359) | |
Polyclonal | |
Unconjugated | |
Ncf1 | |
47 kDa autosomal chronic granulomatous disease protein; 47 kDa neutrophil oxidase factor; NADPH oxidase organizer 2; NADPH oxidase subunit (47 kD); NADPH oxidase subunit (47kDa); Ncf1; NCF-1; NCF1A; NCF-47K; neutrop; neutrophil cytosol factor 1; neutrophil cytosolic factor 1; neutrophil cytosolic factor 1, (chronic granulomatous disease, autosomal 1); neutrophil NADPH oxidase factor 1; Nox organizer 2; NOXO2; nox-organizing protein 2; P47 phox; p47<phox>; p47phox; p47-phox; phagocyte oxidase 47 kDa; SH3 and PX domain-containing protein 1A; SH3PXD1A | |
Rabbit | |
Affinity chromatography | |
RUO | |
653361 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P14598 | |
Ncf1 | |
Peptide sequence around phosphorylation site of Serine 359 (Q-R-S(p)-K-P) derived from Human p47 phox | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |